Search
Search

IL7| Gene Symbol: IL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000880.3 | NP_000871.1 | 6 | 1853 | ||
| NM_001199886.1 | NP_001186815.1 | 5 | 1721 | |||
| NM_001199887.1 | NP_001186816.1 | 5 | 1799 | |||
| NM_001199888.1 | NP_001186817.1 | 4 | 1667 | |||
| XM_017013397.1 | XP_016868886.1 | 6 | 1737 | |||
| GenBank mRNA | BC047698.1 | - | 6 | 1841 | 5748841 | |