Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTRF1 SIRNA Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
MRF1, MTTRF1, RF1
Chromosome Location:
Chr.13: 41216369 - 41311794 on Build GRCh38
UniGene ID:
Hs.382176
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14472
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTRF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004294.2
NP_004285.2
6
941
XM_005266599.2
XP_005266656.1
8
1190
XM_006719897.2
XP_006719960.1
8
1298
XM_006719898.2
XP_006719961.1
8
1282
XM_006719900.2
XP_006719963.1
7
1011
XM_011535315.2
XP_011533617.1
8
1407
XM_011535317.2
XP_011533619.1
7
1290
XM_011535318.2
XP_011533620.1
8
1450
XM_011535320.2
XP_011533622.1
8
1386
XM_011535321.2
XP_011533623.1
11
1522
XM_011535323.2
XP_011533625.1
8
1252
XM_011535325.2
XP_011533627.1
7
1205
XM_011535326.2
XP_011533628.1
7
1251
XM_017020856.1
XP_016876345.1
7
1251
XM_017020857.1
XP_016876346.1
10
1385
XM_017020858.1
XP_016876347.1
9
1380
XM_017020859.1
XP_016876348.1
7
1251
XM_017020860.1
XP_016876349.1
9
1268
XM_017020861.1
XP_016876350.1
9
1305
XM_017020862.1
XP_016876351.1
8
2463
XM_017020863.1
XP_016876352.1
8
2311
XM_017020864.1
XP_016876353.1
8
2203
XM_017020865.1
XP_016876354.1
7
2155
XM_017020866.1
XP_016876355.1
6
2020
XM_017020867.1
XP_016876356.1
8
1168
XM_017020868.1
XP_016876357.1
6
1136
XM_017020869.1
XP_016876358.1
7
1034
XM_017020870.1
XP_016876359.1
8
1298
XM_017020871.1
XP_016876360.1
7
1011
XM_017020872.1
XP_016876361.1
7
1251
XM_017020873.1
XP_016876362.1
11
1592
XM_017020874.1
XP_016876363.1
8
1144
XM_017020875.1
XP_016876364.1
8
1138
XM_017020876.1
XP_016876365.1
7
2478
XM_017020877.1
XP_016876366.1
9
1313
XM_017020878.1
XP_016876367.1
8
1190
GenBank mRNA
AF072934.1
-
6
912
AK055086.1
-
8
1312
AK294343.1
-
7
1098
BC042196.1
-
6
941
5198308
Back To Top
More Information
Panther Classification:
Molecular Function -
translation release factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational termination
mitochondrial translational termination
translation release factor activity
translation release factor activity, codon specific
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0