Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL18 SIRNA Assays ›
          Gene Symbol:
          IL18
          Gene Name:
          interleukin 18
          Gene Aliases:
          IGIF, IL-18, IL-1g, IL1F4
          Chromosome Location:
          Chr.11: 112143251 - 112164117 on Build GRCh38
          UniGene ID:
          Hs.83077
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144778

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243211.1 NP_001230140.1 3 393
          NM_001562.3 NP_001553.1 4 405
          XM_011542805.1 XP_011541107.1 3 401
          XM_011542806.2 XP_011541108.1 4 409
          GenBank mRNA AF077611.1 - 3 185
          AF380360.1 - 4 237
          AY044641.1 - 4 282
          AY266351.1 - 3 350
          BC007007.1 - 4 380 3686124
          BC007461.1 - 4 405 3681009
          CR541973.1 - 3 185
          CR542001.1 - 3 185
          D49950.1 - 4 362
          U90434.1 - 3 182

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine interleukin superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          inflammatory response
          immune response
          cell-cell signaling
          positive regulation of gene expression
          natural killer cell activation
          regulation of cell adhesion
          sleep
          lipopolysaccharide-mediated signaling pathway
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-17 production
          positive regulation of natural killer cell proliferation
          positive regulation of tissue remodeling
          chemokine biosynthetic process
          T-helper 1 type immune response
          type 2 immune response
          interleukin-2 biosynthetic process
          interferon-gamma biosynthetic process
          positive regulation of activated T cell proliferation
          interleukin-13 biosynthetic process
          granulocyte macrophage colony-stimulating factor biosynthetic process
          positive regulation of NF-kappaB import into nucleus
          positive regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of myoblast differentiation
          positive regulation of inflammatory response
          positive regulation of NK T cell proliferation
          cellular response to organic cyclic compound
          cytokine activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0