Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002196.2 | NP_002187.1 | 1 | 2181 | ||
GenBank mRNA | M93119.1 | - | 1 | 2181 |