Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002226.4 | NP_002217.3 | 25 | 3619 | ||
NM_145159.2 | NP_660142.1 | 24 | 3505 | |||
GenBank mRNA | AB209370.1 | - | 1 | 3637 | ||
AF003521.1 | - | 25 | 3247 | |||
AF020201.1 | - | 25 | 3266 | |||
AF029778.1 | - | 25 | 3619 | |||
AF029779.1 | - | 24 | 3505 | |||
LN607838.1 | - | 2 | 368 | |||
Y14330.1 | - | 25 | 3170 |