Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002264.3 | NP_002255.3 | 5 | 541 | ||
NR_026698.1 | 5 | 541 | ||||
XM_005247437.3 | XP_005247494.1 | 5 | 423 | |||
XM_011512795.2 | XP_011511097.1 | 6 | 475 | |||
XM_011512797.2 | XP_011511099.1 | 4 | 358 | |||
XM_017006363.1 | XP_016861852.1 | 5 | 539 | |||
XM_017006364.1 | XP_016861853.1 | 5 | 418 | |||
GenBank mRNA | AK299030.1 | - | 4 | 427 | ||
AK312713.1 | - | 5 | 536 | |||
BC002374.2 | - | 5 | 530 | 2822859 | ||
BC003009.1 | - | 5 | 530 | 2822859 | ||
BC090864.1 | - | 5 | 512 | 6173605 | ||
BT006959.1 | - | 4 | 353 | |||
CR456743.1 | - | 4 | 353 | |||
DA308391.1 | - | 5 | 541 | |||
DN991078.1 | - | 5 | 383 | |||
S75295.1 | - | 5 | 399 | |||
U20620.1 | - | 3 | 206 |