Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK9 SIRNA Assays ›
          Gene Symbol:
          MAPK9
          Gene Name:
          mitogen-activated protein kinase 9
          Gene Aliases:
          JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK
          Chromosome Location:
          Chr.5: 180233594 - 180292071 on Build GRCh38
          UniGene ID:
          Hs.484371
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1452
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135044.1 NP_001128516.1 2, 3 391
          NM_001308244.1 NP_001295173.1 1, 2 167
          NM_002752.4 NP_002743.3 2, 3 391
          NM_139068.2 NP_620707.1 2, 3 391
          NM_139069.2 NP_620708.1 2, 3 391
          NM_139070.2 NP_620709.1 2, 3 391
          XM_006714891.2 XP_006714954.1 3, 4 449
          XM_017009638.1 XP_016865127.1 3, 4 360
          XM_017009641.1 XP_016865130.1 3, 4 458
          XM_017009642.1 XP_016865131.1 2, 3 287
          XM_017009643.1 XP_016865132.1 2, 3 287
          GenBank mRNA AB451302.1 - 1, 2 120
          AB451355.1 - 1, 2 120
          AB451433.1 - 1, 2 120
          AK289638.1 - 2, 3 287
          BC032539.1 - 1, 2 181 5528624
          CR536580.1 - 1, 2 120
          DB451562.1 - 2, 3 417
          EU927388.1 - 1, 2 140
          HM036706.1 - 1, 2 140
          HM036707.1 - 1, 2 140
          HM036708.1 - 1, 2 140
          L31951.1 - 1, 2 178
          U09759.1 - 1, 2 271
          U34821.1 - 1, 2 164
          U35002.1 - 1, 2 142
          U35003.1 - 1, 2 154

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00177102_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytochrome c from mitochondria
          positive regulation of protein phosphorylation
          protein phosphorylation
          protein targeting to mitochondrion
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to stress
          JNK cascade
          JUN phosphorylation
          central nervous system development
          response to mechanical stimulus
          response to toxic substance
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cell morphogenesis involved in differentiation
          response to amine
          neuron projection development
          positive regulation of prostaglandin biosynthetic process
          regulation of protein ubiquitination
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          cellular response to UV
          Fc-epsilon receptor signaling pathway
          response to drug
          regulation of circadian rhythm
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          regulation of JNK cascade
          response to cadmium ion
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          positive regulation of podosome assembly
          positive regulation of apoptotic signaling pathway
          JUN kinase activity
          protein binding
          ATP binding
          transcription factor binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0