Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBPJ SIRNA Assays ›
          Gene Symbol:
          RBPJ
          Gene Name:
          recombination signal binding protein for immunoglobulin kappa J region
          Gene Aliases:
          AOS3, CBF1, IGKJRB, IGKJRB1, KBF2, RBP-J, RBPJK, RBPSUH, SUH, csl
          Chromosome Location:
          Chr.4: 26163489 - 26435131 on Build GRCh38
          UniGene ID:
          Hs.479396
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145253

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBPJ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005349.3 NP_005340.2 6 717
          NM_015874.4 NP_056958.3 5 664
          NM_203283.2 NP_976028.1 5 601
          NM_203284.2 NP_976029.1 6 847
          XM_005248161.3 XP_005248218.1 6 731
          XM_011513840.2 XP_011512142.1 6 1123
          XM_017008170.1 XP_016863659.1 5 520
          XM_017008171.1 XP_016863660.1 5 499
          XM_017008172.1 XP_016863661.1 6 662
          XM_017008173.1 XP_016863662.1 6 940
          XM_017008174.1 XP_016863663.1 6 756
          XM_017008175.1 XP_016863664.1 6 672
          GenBank mRNA AK302230.1 - 5 454
          AK303244.1 - 5 513
          AK311419.1 - 5 500
          BC020780.1 - 5 601 4718839
          BC064976.1 - 6 622 6152924
          BQ216033.1 - 5 645
          L07872.1 - 5 559

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          somitogenesis
          epithelial to mesenchymal transition
          blood vessel remodeling
          inflammatory response to antigenic stimulus
          secondary heart field specification
          outflow tract morphogenesis
          endocardium morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          cardiac left ventricle morphogenesis
          ventricular trabecula myocardium morphogenesis
          regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
          DNA recombination
          transcription initiation from RNA polymerase II promoter
          humoral immune response
          Notch signaling pathway
          positive regulation of transcription of Notch receptor target
          negative regulation of cell proliferation
          auditory receptor cell fate commitment
          epidermal cell fate specification
          pituitary gland development
          B cell differentiation
          keratinocyte differentiation
          negative regulation of ossification
          positive regulation of BMP signaling pathway
          somatic stem cell population maintenance
          dorsal aorta morphogenesis
          atrioventricular canal development
          defense response to bacterium
          myeloid dendritic cell differentiation
          negative regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of timing of cell differentiation
          sebaceous gland development
          hair follicle maturation
          positive regulation of cardiac muscle cell proliferation
          Clara cell differentiation
          labyrinthine layer blood vessel development
          arterial endothelial cell fate commitment
          Notch signaling involved in heart development
          positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
          blood vessel lumenization
          interleukin-4 secretion
          blood vessel endothelial cell fate specification
          positive regulation of ERBB signaling pathway
          positive regulation of ephrin receptor signaling pathway
          positive regulation of canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
          positive regulation of cell proliferation involved in heart morphogenesis
          recombinase activity
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II repressing transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0