Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPH1 SIRNA Assays ›
Gene Symbol:
HNRNPH1
Gene Name:
heterogeneous nuclear ribonucleoprotein H1 (H)
Gene Aliases:
HNRPH, HNRPH1, hnRNPH
Chromosome Location:
Chr.5: 179614178 - 179634784 on Build GRCh38
UniGene ID:
Hs.604001
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145262
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257293.1
NP_001244222.1
14
2141
NM_005520.2
NP_005511.1
14
2111
XM_005265895.2
XP_005265952.1
13
2048
XM_005265896.2
XP_005265953.1
13
2051
XM_005265901.1
XP_005265958.1
12
1913
XM_005265902.4
XP_005265959.1
12
1899
XM_006714862.2
XP_006714925.1
12
1991
XM_006714863.2
XP_006714926.1
13
2041
XM_011534542.1
XP_011532844.1
13
2365
XM_011534544.1
XP_011532846.1
13
2548
XM_011534545.1
XP_011532847.1
13
2256
XM_011534546.1
XP_011532848.1
14
2404
XM_011534547.1
XP_011532849.1
13
2021
XM_017009414.1
XP_016864903.1
12
1988
XM_017009415.1
XP_016864904.1
14
2120
XM_017009416.1
XP_016864905.1
14
2415
XM_017009417.1
XP_016864906.1
15
2454
XM_017009418.1
XP_016864907.1
14
2071
XM_017009419.1
XP_016864908.1
13
2038
XM_017009420.1
XP_016864909.1
11
1852
XM_017009421.1
XP_016864910.1
13
1962
XM_017009422.1
XP_016864911.1
13
1949
XM_017009423.1
XP_016864912.1
13
1932
XM_017009424.1
XP_016864913.1
10
4695
XM_017009425.1
XP_016864914.1
12
1902
GenBank mRNA
AK092273.1
-
13
2306
AK124530.1
-
13
3530
BC001348.2
-
14
2087
2961378
BX647205.1
-
4
5428
L22009.1
-
14
2064
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
RNA processing
fibroblast growth factor receptor signaling pathway
gene expression
regulation of RNA splicing
nucleotide binding
RNA binding
protein binding
poly(U) RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-4vcgr:80/10.239.160.220:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0