Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005546.3 | NP_005537.3 | 17 | 2765 | ||
XM_017009443.1 | XP_016864932.1 | 15 | 2378 | |||
GenBank mRNA | AB209622.1 | - | 16 | 4268 | ||
BX538196.1 | - | 5 | 1505 | |||
D13720.1 | - | 17 | 2765 |