Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000872.4 | NP_000863.1 | 3 | 2093 | ||
NM_019859.3 | NP_062873.1 | 4 | 2191 | |||
NM_019860.3 | NP_062874.1 | 3 | 2098 | |||
GenBank mRNA | BC047526.1 | - | 3 | 2094 | 5297325 | |
BG108256.1 | - | 1 | 551 |
Gene Symbol: HTR7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002774.3 | 1 | 2901 | |||
GenBank mRNA | AK125093.1 | - | 2 | 2762 |
Set Membership: |
G-protein coupled receptor |