Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRIK2 SIRNA Assays ›
          Gene Symbol:
          GRIK2
          Gene Name:
          glutamate ionotropic receptor kainate type subunit 2
          Gene Aliases:
          EAA4, GLR6, GLUK6, GLUR6, GluK2, MRT6
          Chromosome Location:
          Chr.6: 101393708 - 102070083 on Build GRCh38
          UniGene ID:
          Hs.98262
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145383

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GRIK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166247.1 NP_001159719.1 3 813
          NM_021956.4 NP_068775.1 3 813
          NM_175768.3 NP_786944.1 3 813
          XM_005266945.2 XP_005267002.1 4 943
          XM_005266946.3 XP_005267003.1 3 898
          XM_011535777.2 XP_011534079.1 3 980
          XM_011535778.2 XP_011534080.1 3 980
          XM_017010781.1 XP_016866270.1 3 980
          XM_017010782.1 XP_016866271.1 3 980
          GenBank mRNA AJ252246.1 - 3 520
          AJ301610.1 - 3 520
          AK289542.1 - 3 747
          AK291965.1 - 3 769
          AK303721.1 - 3 523
          BC037954.1 - 3 1005
          BC063814.1 - 3 723 6189201
          HM149335.1 - 3 520
          HM149336.1 - 3 520
          HM149337.1 - 3 520
          HM149338.1 - 3 520
          HM149339.1 - 3 520
          U16126.1 - 3 520

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          transport
          cellular calcium ion homeostasis
          intracellular protein transport
          glutamate receptor signaling pathway
          chemical synaptic transmission
          neuronal action potential
          ion transmembrane transport
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          receptor clustering
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          regulation of JNK cascade
          regulation of long-term neuronal synaptic plasticity
          regulation of short-term neuronal synaptic plasticity
          modulation of synaptic transmission
          positive regulation of synaptic transmission
          neuron apoptotic process
          negative regulation of synaptic transmission, glutamatergic
          excitatory postsynaptic potential
          inhibitory postsynaptic potential
          extracellular-glutamate-gated ion channel activity
          ligand-gated ion channel activity
          kainate selective glutamate receptor activity
          PDZ domain binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7d94m:80/100.66.78.9:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0