Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000167.5 | NP_000158.1 | 5 | 543 | ||
NM_001128127.2 | NP_001121599.1 | 5 | 543 | |||
NM_001205019.1 | NP_001191948.1 | 5 | 543 | |||
NM_203391.3 | NP_976325.1 | 5 | 543 | |||
XM_005274488.4 | XP_005274545.1 | 3 | 313 | |||
XM_006724483.2 | XP_006724546.1 | 6 | 580 | |||
XM_006724484.2 | XP_006724547.1 | 6 | 579 | |||
XM_006724485.2 | XP_006724548.1 | 4 | 388 | |||
XM_006724486.3 | XP_006724549.1 | 3 | 373 | |||
XM_011545491.2 | XP_011543793.1 | 6 | 581 | |||
XM_011545492.1 | XP_011543794.1 | 6 | 580 | |||
XM_011545493.2 | XP_011543795.1 | 4 | 263 | |||
XM_011545494.2 | XP_011543796.1 | 4 | 208 | |||
XM_017029409.1 | XP_016884898.1 | 6 | 578 | |||
XM_017029410.1 | XP_016884899.1 | 3 | 357 | |||
XM_017029411.1 | XP_016884900.1 | 4 | 390 | |||
XM_017029412.1 | XP_016884901.1 | 3 | 311 | |||
GenBank mRNA | AK294304.1 | - | 4 | 299 | ||
AK294938.1 | - | 6 | 578 | |||
AK308238.1 | - | 3 | 222 | |||
AK313215.1 | - | 5 | 479 | |||
BC037549.1 | - | 5 | 400 | 5213716 | ||
BC042421.1 | - | 5 | 544 | 4820945 | ||
BC071595.1 | - | 5 | 544 | 5276056 | ||
BM906503.1 | - | 490 | ||||
BX647604.1 | - | 5 | 364 | |||
L13943.1 | - | 5 | 430 | |||
X68285.1 | - | 3 | 136 |