Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC101929829 SIRNA Assays ›
          Gene Symbol:
          LOC101929829 CYP2D6 CYP2D7
          Gene Name:
          cytochrome P450 family 2 subfamily D member 6 pseudogene
          cytochrome P450 family 2 subfamily D member 6
          cytochrome P450 family 2 subfamily D member 7 (gene/pseudogene)
          Gene Aliases:
          -
          CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
          CYP2D, CYP2D7AP, CYP2D7P, CYP2D7P1, CYP2D@, P450C2D, P450DB1, RNA40057
          Chromosome Location:
          -
          Chromosome Location:
          Chr.22: 42125531 - 42130881 on Build GRCh38
          Chromosome Location:
          Chr.22: 42140203 - 42144575 on Build GRCh38
          UniGene ID:
          Hs.648256
          Hs.648256
          Hs.333497
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 145517

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC101929829
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_111920.1 7 1164
          GenBank mRNA AB209492.1 - 6 3343
          AK308211.1 - 6 1349
          M24499.1 - 5 703
          X16865.1 - 5 709
          Gene Symbol: CYP2D6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000106.5 NP_000097.3 7 1165
          NM_001025161.2 NP_001020332.2 6 1012
          XM_011529966.2 XP_011528268.1 7 1159
          XM_011529968.2 XP_011528270.1 7 1159
          XM_011529970.2 XP_011528272.1 6 1006
          XM_011529972.2 XP_011528274.1 6 1017
          GenBank mRNA AK309600.1 - 4 1407
          AY663390.1 - 5 698
          BC066877.1 - 6 923
          BC067432.1 - 7 1076 7002157
          BC075023.2 - 7 1105 30915411
          BC075024.2 - 7 1105 30915622
          BC106757.1 - 6 944 40024713
          BC106758.1 - 7 1097 40024717
          BC126858.1 - 7 1097 40024719
          BX096000.1 - 1 91
          CR456430.1 - 6 947
          M20403.1 - 7 1075
          X07618.1 - 5 966
          X08006.1 - 7 1075
          Gene Symbol: CYP2D7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY220845.1 - 8 1249
          X07619.1 - 4 787
          X07620.1 - 3 167

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          steroid metabolic process
          coumarin metabolic process
          alkaloid metabolic process
          alkaloid catabolic process
          monoterpenoid metabolic process
          drug metabolic process
          arachidonic acid metabolic process
          isoquinoline alkaloid metabolic process
          drug catabolic process
          heterocycle metabolic process
          negative regulation of binding
          oxidation-reduction process
          oxidative demethylation
          negative regulation of cellular organofluorine metabolic process
          monooxygenase activity
          iron ion binding
          drug binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          aromatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0