Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000732.4 | NP_000723.1 | 5 | 689 | ||
NM_001040651.1 | NP_001035741.1 | 4 | 557 | |||
XM_017018543.1 | XP_016874032.1 | 6 | 1589 | |||
GenBank mRNA | BC039035.1 | - | 5 | 553 | 5745355 | |
BC070321.1 | - | 5 | 689 | 30412345 | ||
CD014058.1 | - | 4 | 444 |