Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTNND1 SIRNA Assays ›
          Gene Symbol:
          CTNND1 TMX2-CTNND1
          Gene Name:
          catenin delta 1
          TMX2-CTNND1 readthrough (NMD candidate)
          Gene Aliases:
          CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
          -
          Chromosome Location:
          Chr.11: 57761762 - 57819180 on Build GRCh38
          Chromosome Location:
          Chr.11: 57712523 - 57819180 on Build GRCh38
          UniGene ID:
          Hs.166011
          Hs.166011
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 145775

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTNND1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085458.1 NP_001078927.1 6 1428
          NM_001085459.1 NP_001078928.1 6 1428
          NM_001085460.1 NP_001078929.1 6 1355
          NM_001085461.1 NP_001078930.1 6 1428
          NM_001085462.1 NP_001078931.1 5 1236
          NM_001085463.1 NP_001078932.1 4 947
          NM_001085464.1 NP_001078933.1 4 947
          NM_001085465.1 NP_001078934.1 4 947
          NM_001085466.1 NP_001078935.1 4 947
          NM_001085467.1 NP_001078936.1 4 1020
          NM_001085468.1 NP_001078937.1 4 947
          NM_001085469.1 NP_001078938.1 3 875
          NM_001206883.1 NP_001193812.1 5 1127
          NM_001206884.1 NP_001193813.1 5 1127
          NM_001206885.1 NP_001193814.1 6 1428
          NM_001206886.1 NP_001193815.1 5 1127
          NM_001206887.1 NP_001193816.1 5 1127
          NM_001206888.1 NP_001193817.1 5 1127
          NM_001206889.1 NP_001193818.1 6 1246
          NM_001206890.1 NP_001193819.1 4 969
          NM_001206891.1 NP_001193820.1 6 1319
          NM_001331.2 NP_001322.1 6 1428
          GenBank mRNA AF062317.1 - 6 1393
          AF062319.1 - 5 1092
          AF062320.1 - 5 1092
          AF062321.1 - 6 1393
          AF062322.1 - 5 1092
          AF062323.1 - 4 912
          AF062324.1 - 6 1320
          AF062325.1 - 5 1092
          AF062327.1 - 5 1092
          AF062328.1 - 6 1393
          AF062329.1 - 6 1211
          AF062330.1 - 4 934
          AF062331.1 - 6 1284
          AF062332.1 - 4 912
          AF062334.1 - 4 985
          AF062335.1 - 4 912
          AF062336.1 - 3 840
          AF062337.1 - 4 912
          AF062338.1 - 4 912
          AF062339.1 - 6 1393
          AF062341.1 - 6 1393
          AF062343.1 - 5 1201
          AK292554.1 - 6 1370
          BC010501.1 - 2 549
          BC075795.1 - 6 1244 5269008
          CR749275.1 - 6 1188
          Gene Symbol: TMX2-CTNND1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037646.1 7 1416
          GenBank mRNA AB002382.1 - 7 1322

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell adhesion
          brain development
          Wnt signaling pathway
          single organismal cell-cell adhesion
          adherens junction organization
          negative regulation of canonical Wnt signaling pathway
          receptor binding
          protein binding
          protein kinase binding
          cadherin binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0