Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DAXX SIRNA Assays ›
          Gene Symbol:
          DAXX
          Gene Name:
          death domain associated protein
          Gene Aliases:
          BING2, DAP6, EAP1
          Chromosome Location:
          Chr.6: 33318558 - 33323016 on Build GRCh38
          UniGene ID:
          Hs.336916
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145796

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DAXX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001141969.1 NP_001135441.1 3 495
          NM_001141970.1 NP_001135442.1 3 426
          NM_001254717.1 NP_001241646.1 2 236
          NM_001350.4 NP_001341.1 3 492
          GenBank mRNA AB209493.1 - 1 661
          AF015956.2 - 2 342
          AF039136.1 - 3 435
          AF050179.1 - 3 403
          AF097742.1 - 2 295
          AK223404.1 - 3 493
          AK292187.1 - 3 493
          AK303767.1 - 3 369
          AK303854.1 - 2 193
          AK309612.1 - 2 545
          AK313034.1 - 3 435
          BC109073.1 - 3 378 40034214
          BC109074.1 - 3 378 40034219
          CR457085.1 - 2 288
          DC345308.1 - 3 492
          HQ436528.1 - 2 288
          HQ436529.1 - 2 288

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of protein phosphorylation
          nucleosome assembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          activation of JUN kinase activity
          extrinsic apoptotic signaling pathway via death domain receptors
          response to metal ion
          negative regulation of myotube differentiation
          viral process
          covalent chromatin modification
          androgen receptor signaling pathway
          PML body organization
          regulation of protein ubiquitination
          positive regulation of histone phosphorylation
          positive regulation of apoptotic process
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          cellular response to tumor necrosis factor
          cellular response to anoxia
          positive regulation of neuron death
          p53 binding
          transcription corepressor activity
          receptor signaling protein activity
          protein binding
          transcription factor binding
          enzyme binding
          protein kinase binding
          protein kinase activator activity
          heat shock protein binding
          ubiquitin protein ligase binding
          histone binding
          protein N-terminus binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-89x45:80/100.66.77.245:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0