Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX3X SIRNA Assays ›
          Gene Symbol:
          DDX3X
          Gene Name:
          DEAD-box helicase 3, X-linked
          Gene Aliases:
          CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
          Chromosome Location:
          Chr.X: 41333308 - 41364472 on Build GRCh38
          UniGene ID:
          Hs.728563
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145804

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDX3X
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193416.2 NP_001180345.1 11 1173
          NM_001193417.2 NP_001180346.1 10 1125
          NM_001356.4 NP_001347.3 11 2027
          NR_126093.1 11 2027
          XM_011543892.2 XP_011542194.1 11 1204
          XM_017029313.1 XP_016884802.1 11 3372
          XM_017029314.1 XP_016884803.1 11 3372
          GenBank mRNA AB208983.1 - 11 1125
          AB451220.1 - 11 1082
          AB451343.1 - 11 1082
          AF000982.1 - 11 1938
          AF000983.1 - 11 1938
          AF061337.1 - 11 1157
          AK291153.1 - 11 1173
          AK297159.1 - 11 1173
          AK304661.1 - 10 1130
          AK304689.1 - 10 1125
          AY776161.1 - 3 233
          BC011819.2 - 11 1152 3617040
          U50553.1 - 11 1391

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          chromosome segregation
          extrinsic apoptotic signaling pathway via death domain receptors
          response to virus
          RNA secondary structure unwinding
          positive regulation of gene expression
          Wnt signaling pathway
          negative regulation of translation
          positive regulation of cell growth
          negative regulation of cell growth
          negative regulation of protein complex assembly
          DNA duplex unwinding
          positive regulation of interferon-beta production
          stress granule assembly
          intracellular signal transduction
          mature ribosome assembly
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of viral genome replication
          innate immune response
          positive regulation of translation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of translational initiation
          cellular response to arsenic-containing substance
          cellular response to osmotic stress
          positive regulation of chemokine (C-C motif) ligand 5 production
          intrinsic apoptotic signaling pathway
          cell-cell adhesion
          positive regulation of G1/S transition of mitotic cell cycle
          protein localization to cytoplasmic stress granule
          negative regulation of intrinsic apoptotic signaling pathway
          DNA binding
          RNA binding
          GTPase activity
          ATP-dependent DNA helicase activity
          ATP-dependent RNA helicase activity
          protein binding
          ATP binding
          transcription factor binding
          poly(A) binding
          eukaryotic initiation factor 4E binding
          ATPase activity
          nucleoside-triphosphatase activity
          translation initiation factor binding
          RNA stem-loop binding
          ribosomal small subunit binding
          CTPase activity
          poly(A) RNA binding
          mRNA 5'-UTR binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-v95b6:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0