Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197260.1 | NP_001184189.1 | 5 | 1104 | ||
NM_001318866.1 | NP_001305795.1 | 5 | 1604 | |||
NM_001318867.1 | NP_001305796.1 | 5 | 1649 | |||
NM_001318868.1 | NP_001305797.1 | 4 | 1421 | |||
NM_001318869.1 | NP_001305798.1 | 4 | 1415 | |||
NM_001381.4 | NP_001372.1 | 5 | 1606 | |||
XM_017003528.1 | XP_016859017.1 | 3 | 1527 | |||
GenBank mRNA | AF035299.1 | - | 5 | 1097 | ||
AF180527.1 | - | 4 | 826 | |||
AK055944.1 | - | 5 | 1606 | |||
AK127127.1 | - | 3 | 1302 | |||
AK127656.1 | - | 3 | 1302 | |||
AK222883.1 | - | 5 | 1045 | |||
AK296154.1 | - | 5 | 1059 | |||
AK300097.1 | - | 4 | 826 | |||
AK314678.1 | - | 5 | 1026 | |||
BC111055.1 | - | 4 | 850 | 5752188 | ||
BC114440.1 | - | 5 | 1032 | 40083624 | ||
BC114497.1 | - | 4 | 835 | 40083623 | ||
U70987.1 | - | 5 | 1033 |