Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF009424.1 | - | 6 | 1850 | ||
AF009425.1 | - | 5 | 1796 | |||
AF009426.1 | - | 3 | 1449 | |||
AF009427.1 | - | 2 | 1395 |