Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001419.2 | NP_001410.2 | 6 | 2692 | ||
GenBank mRNA | AI375368.1 | - | 1 | 293 | ||
AL713686.1 | - | 1 | 77 |