Search
Search

CD9| Gene Symbol: CD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001769.3 | NP_001760.1 | 5, 6 | 625 | ||
| XM_011521040.1 | XP_011519342.1 | 5, 6 | 604 | |||
| GenBank mRNA | AB079244.1 | - | 5, 6 | 546 | ||
| AK298431.1 | - | 4, 5 | 255 | |||
| AK315219.1 | - | 5, 6 | 574 | |||
| AY423720.1 | - | 5, 6 | 458 | |||
| AY966455.1 | - | 5, 6 | 552 | |||
| BC011988.1 | - | 5, 6 | 492 | 3860667 | ||
| BM763066.1 | - | 5, 6 | 625 | |||
| L34068.1 | - | 5, 6 | 441 | |||
| M38690.1 | - | 5, 6 | 492 | |||
| X60111.1 | - | 5, 6 | 552 | |||