Search
Search

CD9| Gene Symbol: CD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001769.3 | NP_001760.1 | 6 | 702 | ||
| XM_005253814.3 | XP_005253871.1 | 5 | 847 | |||
| XM_011521040.1 | XP_011519342.1 | 6 | 681 | |||
| GenBank mRNA | AB079244.1 | - | 6 | 623 | ||
| AK296332.1 | - | 5 | 793 | |||
| AK298431.1 | - | 5 | 332 | |||
| AK308700.1 | - | 5 | 695 | |||
| AK309527.1 | - | 2 | 447 | |||
| AK315219.1 | - | 6 | 651 | |||
| AY423720.1 | - | 6 | 535 | |||
| AY966455.1 | - | 6 | 630 | |||
| BC011988.1 | - | 6 | 569 | 3860667 | ||
| L34068.1 | - | 6 | 518 | |||
| M38690.1 | - | 6 | 569 | |||
| X60111.1 | - | 6 | 629 | |||