Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLU SIRNA Assays ›
Gene Symbol:
CLU
Gene Name:
clusterin
Gene Aliases:
AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2
Chromosome Location:
Chr.8: 27596917 - 27616717 on Build GRCh38
UniGene ID:
Hs.436657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146051
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLU
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001831.3
NP_001822.3
9
2700
NR_038335.1
9
2835
NR_045494.1
9
2694
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
microglial cell activation
release of cytochrome c from mitochondria
platelet degranulation
lipid metabolic process
complement activation
complement activation, classical pathway
aging
positive regulation of cell proliferation
response to light stimulus
response to wounding
response to virus
protein import
endocrine pancreas development
central nervous system myelin maintenance
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of protein homooligomerization
positive regulation of tumor necrosis factor production
response to potassium ion
reverse cholesterol transport
estrous cycle
innate immune response
positive regulation of nitric oxide biosynthetic process
positive regulation of cell differentiation
neuron projection morphogenesis
protein stabilization
positive regulation of NF-kappaB transcription factor activity
chaperone-mediated protein complex assembly
response to misfolded protein
chaperone-mediated protein folding
microglial cell proliferation
cellular response to growth factor stimulus
regulation of beta-amyloid clearance
regulation of neuron death
positive regulation of neuron death
positive regulation of beta-amyloid formation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of beta-amyloid formation
regulation of neuronal signal transduction
positive regulation of tau-protein kinase activity
positive regulation of neurofibrillary tangle assembly
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein binding
ATPase activity
ubiquitin protein ligase binding
protein N-terminus binding
chaperone binding
misfolded protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0