Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTGF SIRNA Assays ›
          Gene Symbol:
          CTGF
          Gene Name:
          connective tissue growth factor
          Gene Aliases:
          CCN2, HCS24, IGFBP8, NOV2
          Chromosome Location:
          Chr.6: 131948176 - 131951378 on Build GRCh38
          UniGene ID:
          Hs.410037
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146148

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTGF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001901.2 NP_001892.1 5 1006
          GenBank mRNA AK092280.1 - 5 857
          AK125220.1 - 5 788
          AK290884.1 - 5 1000
          AK315207.1 - 5 1000
          AY395801.1 - 5 923
          AY550024.1 - 5 913
          BC087839.1 - 5 982 6181580
          BT019794.1 - 5 800
          BT019795.1 - 5 800
          CR541734.1 - 5 800
          CR541759.1 - 5 800
          M92934.1 - 5 929
          U14750.1 - 4 580
          X78947.1 - 5 945

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          ossification
          angiogenesis
          regulation of cell growth
          tissue homeostasis
          transcription initiation from RNA polymerase II promoter
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          cell-cell signaling
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          epidermis development
          response to wounding
          response to glucose
          organ senescence
          positive regulation of gene expression
          negative regulation of gene expression
          cell migration
          cell differentiation
          lung development
          regulation of chondrocyte differentiation
          response to estradiol
          positive regulation of collagen biosynthetic process
          response to anoxia
          intracellular signal transduction
          chondrocyte proliferation
          response to amino acid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to peptide hormone
          positive regulation of cell differentiation
          positive regulation of JNK cascade
          positive regulation of cell activation
          response to mineralocorticoid
          positive regulation of stress fiber assembly
          cytosolic calcium ion transport
          positive regulation of cardiac muscle contraction
          negative regulation of cell death
          extracellular matrix constituent secretion
          positive regulation of G0 to G1 transition
          positive regulation of ERK1 and ERK2 cascade
          response to fatty acid
          DNA biosynthetic process
          reactive oxygen species metabolic process
          fibronectin binding
          integrin binding
          protein binding
          insulin-like growth factor binding
          protein C-terminus binding
          growth factor activity
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0