Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001944.2 | NP_001935.2 | 16 | 2898 | ||
XM_011525850.1 | XP_011524152.1 | 16 | 2895 | |||
GenBank mRNA | AK290367.1 | - | 16 | 2898 | ||
BX538327.1 | - | 3 | 1945 | |||
M76482.1 | - | 16 | 2872 |