Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001945.2 | NP_001936.1 | 6 | 1792 | ||
GenBank mRNA | AK222688.1 | - | 7 | 1105 | ||
BC033097.1 | - | 6 | 1779 | 5424240 | ||
M60278.1 | - | 6 | 1778 |