Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG1 SIRNA Assays ›
Gene Symbol:
DLG1
Gene Name:
discs large MAGUK scaffold protein 1
Gene Aliases:
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location:
Chr.3: 197042560 - 197299272 on Build GRCh38
UniGene ID:
Hs.292549
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146326
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098424.1
NP_001091894.1
4
411
NM_001204386.1
NP_001191315.1
4
411
NM_001290983.1
NP_001277912.1
4
480
NM_004087.2
NP_004078.2
4
411
XM_005269289.3
XP_005269346.1
4
693
XM_011512502.2
XP_011510804.1
4
693
XM_011512503.1
XP_011510805.1
4
690
XM_011512505.1
XP_011510807.1
4
690
XM_011512506.1
XP_011510808.1
4
690
XM_011512509.1
XP_011510811.1
4
690
XM_017005800.1
XP_016861289.1
4
828
XM_017005801.1
XP_016861290.1
4
348
XM_017005802.1
XP_016861291.1
4
418
XM_017005803.1
XP_016861292.1
4
339
XM_017005804.1
XP_016861293.1
4
693
XM_017005805.1
XP_016861294.1
4
510
XM_017005806.1
XP_016861295.1
4
690
XM_017005807.1
XP_016861296.1
4
789
XM_017005808.1
XP_016861297.1
4
348
XM_017005809.1
XP_016861298.1
4
450
XM_017005810.1
XP_016861299.1
4
348
XM_017005811.1
XP_016861300.1
4
690
XM_017005812.1
XP_016861301.1
4
344
XM_017005813.1
XP_016861302.1
4
348
XM_017005814.1
XP_016861303.1
4
451
XM_017005815.1
XP_016861304.1
4
690
XM_017005816.1
XP_016861305.1
4
789
XM_017005817.1
XP_016861306.1
4
320
XM_017005818.1
XP_016861307.1
4
422
XM_017005819.1
XP_016861308.1
4
339
XM_017005820.1
XP_016861309.1
4
408
GenBank mRNA
AK293967.1
-
1
68
AK304280.1
-
4
411
AK310596.1
-
4
349
AK310893.1
-
4
695
BC015560.2
-
4
650
BC028486.1
-
4
650
BC140841.1
-
4
376
9021709
BC144651.1
-
4
376
9053182
EF553524.1
-
4
480
U13896.1
-
4
409
U13897.1
-
4
409
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00177739_m1
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
branching involved in ureteric bud morphogenesis
immunological synapse formation
endothelial cell proliferation
lens development in camera-type eye
T cell cytokine production
protein dephosphorylation
actin filament organization
mitotic cell cycle checkpoint
establishment or maintenance of cell polarity
chemical synaptic transmission
nervous system development
positive regulation of cell proliferation
regulation of cell shape
viral process
single organismal cell-cell adhesion
peristalsis
positive regulation of actin filament polymerization
cortical actin cytoskeleton organization
astral microtubule organization
membrane raft organization
regulation of myelination
activation of protein kinase activity
ion transmembrane transport
cellular protein complex localization
T cell activation
negative regulation of T cell proliferation
regulation of membrane potential
amyloid precursor protein metabolic process
receptor clustering
positive regulation of potassium ion transport
cortical microtubule organization
establishment or maintenance of epithelial cell apical/basal polarity
negative regulation of mitotic cell cycle
GMP metabolic process
GDP metabolic process
reproductive structure development
embryonic skeletal system morphogenesis
smooth muscle tissue development
negative regulation of epithelial cell proliferation
establishment of centrosome localization
negative regulation of protein kinase B signaling
hard palate development
negative regulation of ERK1 and ERK2 cascade
bicellular tight junction assembly
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
receptor localization to synapse
regulation of NIK/NF-kappaB signaling
regulation of sodium ion transmembrane transport
negative regulation of p38MAPK cascade
guanylate kinase activity
phosphoprotein phosphatase activity
protein binding
protein C-terminus binding
cytoskeletal protein binding
ligand-gated ion channel activity
potassium channel regulator activity
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase binding
protein complex scaffold
ionotropic glutamate receptor binding
ion channel binding
L27 domain binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0