Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASP3 SIRNA Assays ›
          Gene Symbol:
          CASP3
          Gene Name:
          caspase 3
          Gene Aliases:
          CPP32, CPP32B, SCA-1
          Chromosome Location:
          Chr.4: 184627696 - 184649475 on Build GRCh38
          UniGene ID:
          Hs.141125
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146367

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004346.3 NP_004337.2 8 1247
          NM_032991.2 NP_116786.1 7 1080
          XM_011532301.1 XP_011530603.1 8 1181
          GenBank mRNA AK291337.1 - 8 1218
          BC016926.2 - 8 1199 4419175
          BX647609.1 - 7 1200
          U13737.1 - 8 1208

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          B cell homeostasis
          apoptotic DNA fragmentation
          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular component disassembly involved in execution phase of apoptosis
          cellular response to DNA damage stimulus
          heart development
          sensory perception of sound
          learning or memory
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          response to UV
          response to glucose
          response to X-ray
          regulation of macroautophagy
          protein processing
          hippocampus development
          neuron differentiation
          keratinocyte differentiation
          erythrocyte differentiation
          platelet formation
          negative regulation of B cell proliferation
          response to cobalt ion
          response to estradiol
          response to lipopolysaccharide
          glial cell apoptotic process
          response to tumor necrosis factor
          response to nicotine
          hippo signaling
          wound healing
          response to drug
          response to hydrogen peroxide
          T cell homeostasis
          negative regulation of apoptotic process
          response to amino acid
          positive regulation of neuron apoptotic process
          cell fate commitment
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of activated T cell proliferation
          response to antibiotic
          neurotrophin TRK receptor signaling pathway
          response to glucocorticoid
          neuron apoptotic process
          cellular response to organic cyclic compound
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          execution phase of apoptosis
          protease binding
          aspartic-type endopeptidase activity
          cysteine-type endopeptidase activity
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          death receptor binding
          protein binding
          peptidase activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          phospholipase A2 activator activity
          protein complex binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in execution phase of apoptosis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0