Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DSP SIRNA Assays ›
          Gene Symbol:
          DSP
          Gene Name:
          desmoplakin
          Gene Aliases:
          DCWHKTA, DP
          Chromosome Location:
          Chr.6: 7541575 - 7586717 on Build GRCh38
          UniGene ID:
          Hs.519873
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146455

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DSP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008844.2 NP_001008844.1 1 462
          NM_001319034.1 NP_001305963.1 1 462
          NM_004415.3 NP_004406.2 1 462
          GenBank mRNA AB209992.1 - 1 420
          AB621818.1 - 1 367
          BC140802.1 - 1 395 9021670
          HM151899.1 - 1 400
          M77830.3 - 1 400

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          desmosome organization
          ventricular compact myocardium morphogenesis
          epidermis development
          peptide cross-linking
          keratinocyte differentiation
          adherens junction organization
          wound healing
          intermediate filament organization
          protein localization to adherens junction
          bundle of His cell-Purkinje myocyte adhesion involved in cell communication
          regulation of heart rate by cardiac conduction
          regulation of ventricular cardiac muscle cell action potential
          protein kinase C binding
          structural molecule activity
          structural constituent of cytoskeleton
          protein binding
          protein binding, bridging
          poly(A) RNA binding
          cell adhesion molecule binding
          cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0