Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL2 SIRNA Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
HEL-N1, HELN1, HUB
Chromosome Location:
Chr.9: 23690099 - 23850757 on Build GRCh38
UniGene ID:
Hs.166109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146482
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171195.1
NP_001164666.1
5
848
NM_001171197.1
NP_001164668.1
5
861
NM_004432.3
NP_004423.2
5
867
XM_005251393.3
XP_005251450.1
5
1005
XM_005251394.3
XP_005251451.1
5
1162
XM_005251395.3
XP_005251452.1
5
942
XM_006716734.3
XP_006716797.1
6
1254
XM_006716735.3
XP_006716798.1
6
722
XM_006716736.3
XP_006716799.1
5
1143
XM_011517774.2
XP_011516076.1
6
1268
XM_011517776.2
XP_011516078.1
5
698
XM_011517777.2
XP_011516079.1
6
1254
XM_011517778.2
XP_011516080.2
5
791
XM_011517779.2
XP_011516081.1
6
766
XM_011517780.2
XP_011516082.1
6
1095
XM_011517783.2
XP_011516085.1
6
1128
XM_011517784.2
XP_011516086.1
5
823
XM_011517785.2
XP_011516087.1
5
628
XM_011517786.2
XP_011516088.1
5
1002
XM_017014408.1
XP_016869897.1
7
1269
XM_017014409.1
XP_016869898.1
6
907
XM_017014410.1
XP_016869899.1
6
1051
XM_017014411.1
XP_016869900.1
6
1234
XM_017014412.1
XP_016869901.1
7
1216
XM_017014413.1
XP_016869902.1
7
1215
XM_017014414.1
XP_016869903.1
6
1238
XM_017014415.1
XP_016869904.1
5
684
XM_017014416.1
XP_016869905.1
6
757
XM_017014417.1
XP_016869906.1
6
1128
XM_017014418.1
XP_016869907.1
5
962
XM_017014419.1
XP_016869908.1
5
853
XM_017014420.1
XP_016869909.1
5
856
XM_017014421.1
XP_016869910.1
5
974
XM_017014422.1
XP_016869911.1
5
1143
XM_017014423.1
XP_016869912.1
5
800
XM_017014424.1
XP_016869913.1
5
1146
XM_017014425.1
XP_016869914.1
5
954
XM_017014426.1
XP_016869915.1
4
483
GenBank mRNA
AB209294.1
-
5
867
BC030692.1
-
5
865
4826082
BC042393.1
-
5
851
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
nucleotide binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qlhjl:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0