Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004447.5 | NP_004438.3 | 14 | 1784 | ||
XM_011520605.2 | XP_011518907.1 | 15 | 1844 | |||
GenBank mRNA | AK291777.1 | - | 14 | 1589 | ||
AK292931.1 | - | 14 | 1441 | |||
AK301834.1 | - | 7 | 774 | |||
AK304860.1 | - | 14 | 1784 | |||
AK316134.1 | - | 7 | 693 | |||
AK316239.1 | - | 7 | 966 | |||
BC030010.2 | - | 14 | 1596 | 4820933 | ||
U12535.1 | - | 14 | 1556 |