Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006729.4 | NP_006720.1 | 3 | 677 | ||
NM_007309.3 | NP_009293.1 | 3 | 677 | |||
GenBank mRNA | AK291272.1 | - | 3 | 559 | ||
BC117414.1 | - | 3 | 323 | 40125965 | ||
BC143838.1 | - | 3 | 323 | 9052353 | ||
BX363708.2 | - | 3 | 676 | |||
Y15908.1 | - | 3 | 631 | |||
Y15909.1 | - | 3 | 631 |