Search
Search

CAMK2A| Gene Symbol: CAMK2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015981.3 | NP_057065.2 | 19 | 3734 | ||
| NM_171825.2 | NP_741960.1 | 18 | 3701 | |||
| GenBank mRNA | AB023185.1 | - | 18 | 3637 | ||
| BC040457.1 | - | 18 | 3625 | 4811948 | ||