Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ECT2 SIRNA Assays ›
Gene Symbol:
ECT2
Gene Name:
epithelial cell transforming 2
Gene Aliases:
ARHGEF31
Chromosome Location:
Chr.3: 172750682 - 172829273 on Build GRCh38
UniGene ID:
Hs.518299
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146756
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258315.1
NP_001245244.1
18, 19
2099
NM_001258316.1
NP_001245245.1
18, 19
2329
NM_018098.5
NP_060568.3
17, 18
2006
XM_005247176.1
XP_005247233.1
17, 18
2009
XM_006713523.3
XP_006713586.1
18, 19
2088
XM_006713524.3
XP_006713587.1
18, 19
2086
XM_011512514.2
XP_011510816.1
19, 20
2412
XM_011512515.2
XP_011510817.1
18, 19
2068
XM_011512516.1
XP_011510818.1
17, 18
1970
XM_017005830.1
XP_016861319.1
19, 20
2376
XM_017005831.1
XP_016861320.1
18, 19
2003
XM_017005832.1
XP_016861321.1
18, 19
2317
XM_017005833.1
XP_016861322.1
18, 19
2303
XM_017005834.1
XP_016861323.1
19, 20
2409
XM_017005835.1
XP_016861324.1
19, 20
2350
XM_017005836.1
XP_016861325.1
18, 19
2037
XM_017005837.1
XP_016861326.1
17, 18
1922
XM_017005838.1
XP_016861327.1
17, 18
1883
GenBank mRNA
AK023267.1
-
16, 17
1839
AK027713.1
-
7, 8
763
AK308391.1
-
18, 19
2021
AK314581.1
-
17, 18
1950
AY376439.1
-
18, 19
2252
BC006838.1
-
3, 4
221
BC112086.1
-
16, 17
1885
8327554
DQ847274.1
-
17, 18
1904
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
cytokinesis
protein transport
activation of protein kinase activity
positive regulation of cytokinesis
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of protein import into nucleus
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of GTPase activity
positive regulation of neuron differentiation
regulation of protein kinase activity
regulation of small GTPase mediated signal transduction
protein homooligomerization
regulation of attachment of spindle microtubules to kinetochore
cellular response to hydrogen peroxide
bicellular tight junction assembly
cellular response to calcium ion
cellular response to ionizing radiation
activation of GTPase activity
signal transducer activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
Rho GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0