Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ECT2 SIRNA Assays ›
          Gene Symbol:
          ECT2
          Gene Name:
          epithelial cell transforming 2
          Gene Aliases:
          ARHGEF31
          Chromosome Location:
          Chr.3: 172750682 - 172829273 on Build GRCh38
          UniGene ID:
          Hs.518299
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146757

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ECT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258315.1 NP_001245244.1 23, 24 2711
          NM_001258316.1 NP_001245245.1 23, 24 2941
          NM_018098.5 NP_060568.3 22, 23 2618
          XM_005247176.1 XP_005247233.1 22, 23 2621
          XM_006713523.3 XP_006713586.1 23, 24 2700
          XM_006713524.3 XP_006713587.1 23, 24 2698
          XM_011512514.2 XP_011510816.1 24, 25 3024
          XM_011512515.2 XP_011510817.1 23, 24 2680
          XM_011512516.1 XP_011510818.1 22, 23 2582
          XM_017005830.1 XP_016861319.1 24, 25 2988
          XM_017005831.1 XP_016861320.1 23, 24 2615
          XM_017005832.1 XP_016861321.1 23, 24 2929
          XM_017005833.1 XP_016861322.1 23, 24 2915
          XM_017005834.1 XP_016861323.1 24, 25 3021
          XM_017005835.1 XP_016861324.1 24, 25 2962
          XM_017005836.1 XP_016861325.1 23, 24 2649
          XM_017005837.1 XP_016861326.1 22, 23 2534
          XM_017005838.1 XP_016861327.1 22, 23 2495
          GenBank mRNA AK001323.1 - 4, 5 519
          AK027713.1 - 12, 13 1375
          AK308391.1 - 23, 24 2634
          AK314581.1 - 22, 23 2562
          AL137710.1 - 4, 5 534
          AY376439.1 - 23, 24 2864
          BC006838.1 - 8, 9 833
          BC112086.1 - 21, 22 2497 8327554
          DQ847274.1 - 22, 23 2516

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          cytokinesis
          protein transport
          activation of protein kinase activity
          positive regulation of cytokinesis
          regulation of Rho protein signal transduction
          intracellular signal transduction
          positive regulation of protein import into nucleus
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of GTPase activity
          positive regulation of neuron differentiation
          regulation of protein kinase activity
          regulation of small GTPase mediated signal transduction
          protein homooligomerization
          regulation of attachment of spindle microtubules to kinetochore
          cellular response to hydrogen peroxide
          bicellular tight junction assembly
          cellular response to calcium ion
          cellular response to ionizing radiation
          activation of GTPase activity
          signal transducer activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity
          protein binding
          Rho GTPase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0