Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ELAVL4 SIRNA Assays ›
          Gene Symbol:
          ELAVL4
          Gene Name:
          ELAV like neuron-specific RNA binding protein 4
          Gene Aliases:
          HUD, PNEM
          Chromosome Location:
          Chr.1: 50048014 - 50203786 on Build GRCh38
          UniGene ID:
          Hs.213050
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146783

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ELAVL4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144774.2 NP_001138246.1 3 585
          NM_001144775.2 NP_001138247.2 3 435
          NM_001144776.2 NP_001138248.1 3 397
          NM_001144777.2 NP_001138249.1 3 475
          NM_001294348.1 NP_001281277.1 3 655
          NM_001324208.1 NP_001311137.1 3 475
          NM_001324209.1 NP_001311138.1 4 672
          NM_001324212.1 NP_001311141.1 4 672
          NM_001324213.1 NP_001311142.1 3 655
          NM_001324214.1 NP_001311143.1 4 742
          NM_001324215.1 NP_001311144.1 3 655
          NM_001324216.1 NP_001311145.1 3 585
          NM_001324217.1 NP_001311146.1 3 655
          NM_021952.4 NP_068771.2 3 585
          NR_136725.1 4 742
          XM_006710411.3 XP_006710474.1 3 567
          XM_011540889.2 XP_011539191.1 3 450
          XM_011540890.2 XP_011539192.1 3 449
          XM_011540893.2 XP_011539195.1 3 397
          XM_011540894.2 XP_011539196.1 4 739
          XM_011540895.2 XP_011539197.1 4 643
          XM_017000538.1 XP_016856027.1 3 569
          XM_017000539.1 XP_016856028.1 3 1196
          XM_017000540.1 XP_016856029.1 3 453
          XM_017000541.1 XP_016856030.1 3 585
          XM_017000542.1 XP_016856031.1 3 434
          GenBank mRNA AK297338.1 - 3 475
          AK298817.1 - 3 464
          AK298902.1 - 3 655
          AK309653.1 - 3 430
          AY033995.1 - 3 397
          AY033996.1 - 3 408
          AY033997.1 - 3 572
          AY033998.1 - 3 539
          BC036071.1 - 3 543 5286347
          BM509527.1 - 3 442
          DC351092.1 - 3 475
          M62843.1 - 3 409

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001077651, XM_006238593, XM_006238592, XM_006238591, XM_006238595, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001163399, NM_001163397, NM_010488, NM_001038698, AK079088, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA processing
          mRNA processing
          nucleotide binding
          RNA binding
          mRNA 3'-UTR binding
          AU-rich element binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7mgp5:80/100.66.78.208:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0