Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTN1 SIRNA Assays ›
Gene Symbol:
ACTN1
Gene Name:
actinin alpha 1
Gene Aliases:
BDPLT15
Chromosome Location:
Chr.14: 68874123 - 68979366 on Build GRCh38
UniGene ID:
Hs.235750
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147018
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102.3
NP_001093.1
13
1707
NM_001130004.1
NP_001123476.1
13
1707
NM_001130005.1
NP_001123477.1
13
1707
XM_011537265.2
XP_011535567.1
13
1523
XM_011537266.2
XP_011535568.1
13
1523
XM_011537267.2
XP_011535569.1
13
1499
XM_011537268.2
XP_011535570.1
13
1499
XM_017021720.1
XP_016877209.1
13
1523
XM_017021721.1
XP_016877210.1
13
1499
XM_017021722.1
XP_016877211.1
13
1668
XM_017021723.1
XP_016877212.1
13
1644
XM_017021725.1
XP_016877214.1
13
1842
XM_017021726.1
XP_016877215.1
13
1818
XM_017021727.1
XP_016877216.1
13
1668
XM_017021728.1
XP_016877217.1
13
1818
GenBank mRNA
AK098203.1
-
5
440
AK225782.1
-
13
1604
AK294317.1
-
7
910
AK295099.1
-
13
1603
AK298496.1
-
13
1482
AK299362.1
-
13
1345
BC003576.1
-
13
1639
3547017
BC015766.1
-
13
1514
4856734
BT007207.1
-
13
1397
BX248020.1
-
13
1370
BX641076.1
-
2
2849
DQ496098.1
-
13
1580
EU716325.1
-
13
1417
FJ410030.1
-
13
1417
M95178.1
-
13
1508
X15804.1
-
13
1595
X55187.1
-
4
509
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031005, AY437436, BC074001, AF115386.
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
actin filament organization
platelet activation
platelet formation
platelet morphogenesis
regulation of apoptotic process
focal adhesion assembly
actin filament bundle assembly
negative regulation of cellular component movement
actin filament network formation
actin crosslink formation
platelet aggregation
regulation of nucleic acid-templated transcription
double-stranded RNA binding
integrin binding
calcium ion binding
protein binding
vinculin binding
ligand-dependent nuclear receptor transcription coactivator activity
protein homodimerization activity
ion channel binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0