Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB1 SIRNA Assays ›
Gene Symbol:
APBB1
Gene Name:
amyloid beta precursor protein binding family B member 1
Gene Aliases:
FE65, MGC:9072, RIR
Chromosome Location:
Chr.11: 6395124 - 6419830 on Build GRCh38
UniGene ID:
Hs.372840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147078
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164.4
NP_001155.1
11
1706
NM_001257319.2
NP_001244248.1
10
1033
NM_001257320.2
NP_001244249.1
10
1121
NM_001257321.2
NP_001244250.1
11
1169
NM_001257323.2
NP_001244252.1
9
1027
NM_001257325.2
NP_001244254.1
10
929
NM_001257326.2
NP_001244255.1
10
1129
NM_145689.2
NP_663722.1
10
1700
NR_047512.2
11
1706
XM_006718209.3
XP_006718272.2
10
2044
XM_006718210.3
XP_006718273.2
10
2079
XM_006718211.1
XP_006718274.1
10
2250
XM_006718212.3
XP_006718275.2
11
1756
XM_006718213.1
XP_006718276.1
10
2235
XM_006718214.3
XP_006718277.2
10
1750
XM_006718215.3
XP_006718278.3
11
2064
XM_011520037.2
XP_011518339.1
9
1730
XM_011520038.2
XP_011518340.1
9
1235
XM_011520040.2
XP_011518342.1
10
944
XM_011520042.1
XP_011518344.1
9
1225
XM_017017639.1
XP_016873128.1
11
1962
XM_017017640.1
XP_016873129.1
10
1956
XM_017017641.1
XP_016873130.1
10
1941
XM_017017642.1
XP_016873131.1
9
1436
GenBank mRNA
AF394214.1
-
11
1665
AK293452.1
-
12
1210
AK293550.1
-
10
1127
AK293554.1
-
10
897
AK293613.1
-
10
761
AK293643.1
-
10
1121
AK293711.1
-
9
740
AK294495.1
-
9
1075
AK295241.1
-
9
992
AK295425.1
-
10
918
AK295837.1
-
9
1107
AK297542.1
-
4
616
AK297544.1
-
12
1233
AK297550.1
-
11
1170
AK311101.1
-
10
1132
AK311141.1
-
10
962
AK311149.1
-
10
962
AK315965.1
-
10
1120
BC010854.2
-
10
1654
3905603
BX538185.1
-
10
943
EF103274.2
-
10
977
L77864.1
-
11
1665
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_080478, XM_006229935, XM_006229941, XM_006229940, XM_008759717, and others.
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
signal transduction
axonogenesis
response to iron ion
positive regulation of neuron projection development
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone binding
tau protein binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0