Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEPT2 SIRNA Assays ›
          Gene Symbol:
          SEPT2
          Gene Name:
          septin 2
          Gene Aliases:
          DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
          Chromosome Location:
          Chr.2: 241315187 - 241354027 on Build GRCh38
          UniGene ID:
          Hs.721234
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 14709

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEPT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008491.2 NP_001008491.1 8 1057
          NM_001008492.2 NP_001008492.1 8 739
          NM_001282972.1 NP_001269901.1 8 788
          NM_001282973.1 NP_001269902.1 11 1012
          NM_001321029.1 NP_001307958.1 8 773
          NM_001321030.1 NP_001307959.1 8 771
          NM_001321031.1 NP_001307960.1 9 874
          NM_001321032.1 NP_001307961.1 7 654
          NM_001321033.1 NP_001307962.1 9 890
          NM_001321034.1 NP_001307963.1 9 889
          NM_001321035.1 NP_001307964.1 8 755
          NM_004404.4 NP_004395.1 7 654
          NM_006155.2 NP_006146.1 7 956
          XM_011511237.1 XP_011509539.1 7 620
          XM_011511238.1 XP_011509540.1 6 488
          XM_011511240.1 XP_011509542.1 6 497
          XM_017004189.1 XP_016859678.1 9 849
          XM_017004190.1 XP_016859679.1 9 784
          XM_017004191.1 XP_016859680.1 9 849
          XM_017004192.1 XP_016859681.1 9 849
          XM_017004193.1 XP_016859682.1 7 654
          XM_017004194.1 XP_016859683.1 9 804
          XM_017004195.1 XP_016859684.1 9 1008
          XM_017004196.1 XP_016859685.1 9 890
          XM_017004197.1 XP_016859686.1 8 705
          XM_017004198.1 XP_016859687.1 8 767
          XM_017004199.1 XP_016859688.1 9 806
          XM_017004200.1 XP_016859689.1 8 769
          XM_017004201.1 XP_016859690.1 8 891
          XM_017004202.1 XP_016859691.1 7 873
          XM_017004203.1 XP_016859692.1 8 697
          XM_017004204.1 XP_016859693.1 7 612
          XM_017004205.1 XP_016859694.1 9 804
          XM_017004206.1 XP_016859695.1 8 705
          XM_017004207.1 XP_016859696.1 5 1103
          XM_017004208.1 XP_016859697.1 8 1313
          XM_017004209.1 XP_016859698.1 7 1200
          XM_017004210.1 XP_016859699.1 7 1244
          XM_017004211.1 XP_016859700.1 6 1097
          XM_017004212.1 XP_016859701.1 6 1349
          GenBank mRNA AF038404.1 - 7 593
          AK294563.1 - 8 696
          BC014455.1 - 8 936 4906575
          BC033559.1 - 11 990 4821654
          BX648000.1 - 8 673
          D28540.1 - 6 577
          D63878.1 - 7 773

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of L-glutamate transport
          mitotic nuclear division
          smoothened signaling pathway
          neuron projection development
          regulation of protein localization
          cilium assembly
          regulation of catalytic activity
          cell division
          cell-cell adhesion
          protein binding
          GTP binding
          enzyme regulator activity
          protein complex scaffold
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0