Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004421.2 | NP_004412.2 | 2 | 238 | ||
XM_005244731.3 | XP_005244788.1 | 2 | 268 | |||
XM_005244732.3 | XP_005244789.1 | 2 | 268 | |||
XM_005244733.3 | XP_005244790.1 | 2 | 267 | |||
GenBank mRNA | AF006011.1 | - | 2 | 194 | ||
AK293286.1 | - | 2 | 44 | |||
BC050454.1 | - | 2 | 261 | 6201003 | ||
U46461.1 | - | 2 | 335 |