Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004085.3 | NP_004076.1 | 2 | 1364 | ||
GenBank mRNA | BC006994.1 | - | 2 | 1114 | 4051368 | |
BC015093.1 | - | 2 | 1117 | 3684305 | ||
BC070284.1 | - | 2 | 1097 | 4281013 | ||
U66035.1 | - | 2 | 1094 |