Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ACSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004457.3 | NP_004448.2 | 4, 5 | 837 | ||
NM_203372.1 | NP_976251.1 | 3, 4 | 730 | |||
XM_017003584.1 | XP_016859073.1 | 2, 3 | 649 | |||
GenBank mRNA | AB061712.1 | - | 4, 5 | 805 | ||
AK023191.1 | - | 1, 2 | 382 | |||
AK314621.1 | - | 4, 5 | 815 | |||
BC041692.1 | - | 4, 5 | 829 | 5226905 | ||
D89053.1 | - | 2, 3 | 528 |