Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 2 | 673 | ||
GenBank mRNA | AB084459.1 | - | 2 | 516 | ||
AB209179.1 | - | 1 | 2532 | |||
AK303263.1 | - | 3 | 289 | |||
AK308276.1 | - | 3 | 531 | |||
AK313227.1 | - | 2 | 597 | |||
BC002369.2 | - | 2 | 565 | 2822226 | ||
BC003002.1 | - | 2 | 543 | 2823623 | ||
BC014846.1 | - | 2 | 518 | 3854860 | ||
L19559.1 | - | 2 | 575 | |||
U01038.1 | - | 2 | 575 | |||
X73458.1 | - | 2 | 589 | |||
X75932.1 | - | 2 | 575 |