Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007277.4 | NP_009208.2 | 4 | 928 | ||
XM_011513948.2 | XP_011512250.1 | 4 | 963 | |||
XM_011513949.2 | XP_011512251.1 | 4 | 980 | |||
XM_011513950.2 | XP_011512252.1 | 3 | 858 | |||
XM_011513951.2 | XP_011512253.1 | 4 | 957 | |||
XM_017009000.1 | XP_016864489.1 | 4 | 963 | |||
XM_017009001.1 | XP_016864490.1 | 4 | 963 | |||
XM_017009002.1 | XP_016864491.1 | 4 | 963 | |||
XM_017009003.1 | XP_016864492.1 | 4 | 963 | |||
XM_017009004.1 | XP_016864493.1 | 5 | 1041 | |||
XM_017009005.1 | XP_016864494.1 | 4 | 957 | |||
XM_017009006.1 | XP_016864495.1 | 4 | 957 | |||
GenBank mRNA | AK074086.1 | - | 3 | 919 | ||
AK131103.1 | - | 1 | 409 | |||
AL832434.1 | - | 2 | 584 | |||
BC001511.2 | - | 1 | 340 | |||
BC019304.1 | - | 2 | 510 | |||
BC064569.1 | - | 3 | 870 | 5590332 | ||
GU727644.1 | - | 4 | 928 |