Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABL1 SIRNA Assays ›
          Gene Symbol:
          ABL1
          Gene Name:
          ABL proto-oncogene 1, non-receptor tyrosine kinase
          Gene Aliases:
          ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
          Chromosome Location:
          Chr.9: 130713881 - 130887675 on Build GRCh38
          UniGene ID:
          Hs.431048
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147449

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005157.5 NP_005148.2 11 5373
          NM_007313.2 NP_009297.2 11 5677
          GenBank mRNA AA524892.1 - 1 75
          AB209456.1 - 11 5383
          AB209642.1 - 11 5605
          X16416.1 - 11 5327
          X51945.1 - 1 2168

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          B-1 B cell homeostasis
          positive regulation of protein phosphorylation
          B cell proliferation involved in immune response
          transitional one stage B cell differentiation
          mismatch repair
          regulation of transcription, DNA-templated
          cellular protein modification process
          autophagy
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          response to oxidative stress
          cell cycle arrest
          mitotic nuclear division
          epidermal growth factor receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          intrinsic apoptotic signaling pathway in response to DNA damage
          regulation of autophagy
          peptidyl-tyrosine phosphorylation
          cerebellum morphogenesis
          negative regulation of cell-cell adhesion
          microspike assembly
          actin cytoskeleton organization
          regulation of endocytosis
          regulation of cell adhesion
          negative regulation of BMP signaling pathway
          regulation of axon extension
          regulation of microtubule polymerization
          regulation of actin cytoskeleton organization
          positive regulation of osteoblast proliferation
          substrate adhesion-dependent cell spreading
          cellular response to oxidative stress
          platelet-derived growth factor receptor-beta signaling pathway
          peptidyl-tyrosine autophosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          regulation of cell proliferation
          signal transduction in response to DNA damage
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          establishment of protein localization
          negative regulation of mitotic cell cycle
          positive regulation of mitotic cell cycle
          alpha-beta T cell differentiation
          protein autophosphorylation
          spleen development
          thymus development
          collateral sprouting
          positive regulation of peptidyl-tyrosine phosphorylation
          activated T cell proliferation
          B cell receptor signaling pathway
          neuromuscular process controlling balance
          positive regulation of muscle cell differentiation
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of oxidoreductase activity
          negative regulation of ubiquitin-protein transferase activity
          mitochondrial depolarization
          Bergmann glial cell differentiation
          cellular response to hydrogen peroxide
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          negative regulation of protein serine/threonine kinase activity
          actin filament branching
          positive regulation of interleukin-2 secretion
          negative regulation of phospholipase C activity
          positive regulation of neuron death
          positive regulation of interferon-gamma secretion
          regulation of extracellular matrix organization
          cellular response to dopamine
          positive regulation of microtubule binding
          positive regulation of actin filament binding
          activation of protein kinase C activity
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          regulation of cell motility
          regulation of actin cytoskeleton reorganization
          negative regulation of endothelial cell apoptotic process
          negative regulation of cellular senescence
          regulation of response to DNA damage stimulus
          magnesium ion binding
          DNA binding
          actin monomer binding
          nicotinate-nucleotide adenylyltransferase activity
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein kinase C binding
          receptor binding
          protein binding
          ATP binding
          protein C-terminus binding
          SH3 domain binding
          syntaxin binding
          manganese ion binding
          actin filament binding
          mitogen-activated protein kinase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0