Search
Search

COMMD3
COMMD3-BMI1| Gene Symbol: COMMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012071.3 | NP_036203.1 | 3 | 328 | ||
| GenBank mRNA | AF078848.1 | - | 3 | 303 | ||
| AF201948.1 | - | 3 | 322 | |||
| AK128789.1 | - | 3 | 302 | |||
| AY542159.1 | - | 3 | 293 | |||
| BC022898.2 | - | 3 | 315 | 3531636 | ||
| BF695081.1 | - | 329 | ||||
| BG496810.1 | - | 3 | 322 | |||
| BX647275.1 | - | 3 | 325 | |||
| Gene Symbol: COMMD3-BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204062.1 | NP_001190991.1 | 3 | 328 | ||
| GenBank mRNA | AK313235.1 | - | 3 | 303 | ||