Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042681.1 | NP_001036146.1 | 5 | 1230 | ||
NM_012102.3 | NP_036234.3 | 6 | 1415 | |||
XM_005263464.2 | XP_005263521.1 | 5 | 1169 | |||
XM_011541510.1 | XP_011539812.1 | 4 | 1155 | |||
XM_011541511.1 | XP_011539813.1 | 5 | 1281 | |||
XM_017001358.1 | XP_016856847.1 | 5 | 920 | |||
XM_017001359.1 | XP_016856848.1 | 5 | 1425 | |||
GenBank mRNA | AB007927.2 | - | 4 | 565 | ||
AB036737.1 | - | 6 | 1241 | |||
AK096094.1 | - | 5 | 1157 |