Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MED24 SIRNA Assays ›
Gene Symbol:
MED24
Gene Name:
mediator complex subunit 24
Gene Aliases:
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
Chromosome Location:
Chr.17: 40019097 - 40054636 on Build GRCh38
UniGene ID:
Hs.462983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147534
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MED24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079518.1
NP_001072986.1
11, 12
1444
NM_001267797.1
NP_001254726.1
11, 12
1422
NM_014815.3
NP_055630.2
12, 13
1461
NR_052017.1
11, 12
1387
XM_005257874.2
XP_005257931.1
12, 13
1366
XM_006722204.2
XP_006722267.1
12, 13
1366
XM_006722206.2
XP_006722269.1
12, 13
1274
XM_006722207.2
XP_006722270.1
12, 13
1242
XM_011525529.2
XP_011523831.1
13, 14
1423
XM_011525530.2
XP_011523832.1
13, 14
1299
XM_011525531.2
XP_011523833.1
12, 13
1242
XM_011525532.2
XP_011523834.1
13, 14
1331
XM_011525533.2
XP_011523835.1
13, 14
1299
XM_011525534.2
XP_011523836.1
12, 13
1242
XM_011525535.2
XP_011523837.1
12, 13
1242
XM_011525536.2
XP_011523838.1
13, 14
1299
XM_011525538.2
XP_011523840.1
12, 13
1557
XM_017025458.1
XP_016880947.1
13, 14
1331
XM_017025459.1
XP_016880948.1
12, 13
1274
XM_017025460.1
XP_016880949.1
13, 14
1331
XM_017025461.1
XP_016880950.1
13, 14
1423
XM_017025462.1
XP_016880951.1
13, 14
1331
XM_017025463.1
XP_016880952.1
13, 14
1299
XM_017025464.1
XP_016880953.1
12, 13
1242
XM_017025465.1
XP_016880954.1
12, 13
1366
XM_017025466.1
XP_016880955.1
12, 13
1274
XM_017025467.1
XP_016880956.1
12, 13
1274
XM_017025468.1
XP_016880957.1
12, 13
1242
XM_017025469.1
XP_016880958.1
12, 13
1557
XM_017025470.1
XP_016880959.1
12, 13
1557
XM_017025471.1
XP_016880960.1
12, 13
1242
GenBank mRNA
AF055995.1
-
11, 12
1148
AF277379.1
-
11, 12
1148
AK022508.1
-
11, 12
1213
AK291040.1
-
11, 12
1193
AK293309.1
-
10, 11
983
AK299864.1
-
12, 13
1078
AK300992.1
-
11, 12
1066
AK303745.1
-
11, 12
1157
BC011375.1
-
11, 12
1423
3866038
D50920.1
-
12, 13
1221
EU392550.1
-
5, 6
326
Back To Top
More Information
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
protein ubiquitination
histone acetylation
stem cell population maintenance
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
positive regulation of transcription, DNA-templated
protein heterooligomerization
RNA polymerase II transcription cofactor activity
transcription cofactor activity
histone acetyltransferase activity
receptor activity
protein binding
ligand-dependent nuclear receptor transcription coactivator activity
vitamin D receptor binding
thyroid hormone receptor binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0