Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PHLDB1 SIRNA Assays ›
          Gene Symbol:
          PHLDB1
          Gene Name:
          pleckstrin homology like domain family B member 1
          Gene Aliases:
          LL5A
          Chromosome Location:
          Chr.11: 118606296 - 118658038 on Build GRCh38
          UniGene ID:
          Hs.504062
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147559

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PHLDB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144758.2 NP_001138230.1 15, 16 3258
          NM_001144759.2 NP_001138231.1 14, 15 3117
          NM_015157.3 NP_055972.1 16, 17 3481
          XM_005271470.3 XP_005271527.1 15, 16 4027
          XM_005271471.3 XP_005271528.1 15, 16 4027
          XM_005271473.3 XP_005271530.1 14, 15 3886
          XM_005271474.4 XP_005271531.1 14, 15 3886
          XM_006718799.3 XP_006718862.1 16, 17 4156
          XM_006718800.3 XP_006718863.1 16, 17 4156
          XM_006718802.3 XP_006718865.1 15, 16 4015
          XM_006718803.3 XP_006718866.1 15, 16 4015
          XM_011542705.2 XP_011541007.1 18, 19 4471
          XM_011542706.2 XP_011541008.1 18, 19 4471
          XM_011542715.2 XP_011541017.1 9, 10 2393
          XM_017017408.1 XP_016872897.1 17, 18 4327
          XM_017017409.1 XP_016872898.1 17, 18 4300
          GenBank mRNA AB014538.2 - 15, 16 3235
          AB094090.1 - 16, 17 3481
          AK027510.1 - 6, 7 746
          AK074070.2 - 14, 15 3066
          AK074918.1 - 8, 9 953
          AK125783.1 - 17, 18 2485
          AK296345.1 - 4, 5 626
          AL832373.1 - 14, 15 3074
          BC013031.2 - 8, 9 1103
          BX648176.1 - 9, 10 1269

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          regulation of gastrulation
          regulation of epithelial to mesenchymal transition
          regulation of microtubule cytoskeleton organization
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ghg98:80/100.66.77.96:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0